Enghalsflaschen, Umweltproben, mit Schraubverschlüssen, Nalgene®
Lieferant: Thermo Fisher Scientific
332089-0004 332089-0008 332089-0016 332089-0032
331-4241EA 753 CHF
331-4241 331-4242 331-4243 331-4244
Enghalsflaschen, Umweltproben, mit Schraubverschlüssen, Nalgene®
Flaschen Kunststoffflaschen
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant from −100 to +120 °C
  • Leakproof
Narrow neck economy bottles are lightweight versions of the original Nalgene® Lab Quality Bottle, suitable for lighter-duty, general purpose applications.