3-Bromanisol

Lieferant: Molekula

Synonyms: 1-Brom-3-methoxybenzol

MOLE42501045-100GEA 0 CHF
MOLE42501045-100G
3-Bromanisol
3-Bromanisol
Formel: C₇H₇BrO
MW: 187,04 g/mol
Siedepunkt: 211 °C (1013 hPa)
Melting Pt: 2 °C
Dichte: 1,49 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000081
CAS-Nummer: 2398-37-0
EINECS: 219-264-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR