Ethylcinnamat

Lieferant: Roth Carl
Achtung

Synonyms: 3-PhenylacrylsäureethylesterZimtsäureethylesterCinnamsäureethylester3-Phenylpropensäureethylester

ROTH3211.1EA 0 CHF
ROTH3211.1
Ethylcinnamat
Ethylcinnamat
Formel: C₁₁H₁₂O₂
MW: 176,21 g/mol
Siedepunkt: 271 °C (1013 hPa)
Melting Pt: 7 °C
Dichte: 1,046 g/cm³ (20 °C)
MDL Number: MFCD00009189
CAS-Nummer: 103-36-6
EINECS: 203-104-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR