Dapson

Lieferant: Molekula
Achtung

Synonyms: DDS4,4'-Diaminodiphenylsulfon4,4'-Sulfonyldianilin4-AminophenylsulfonBis(4-aminophenyl)sulfon

MOLEM87192589EA 0 CHF
MOLEM87192589
Dapson
Dapson
Formel: C₁₂H₁₂N₂O₂S
MW: 248,31 g/mol
MDL Number: MFCD00007887
CAS-Nummer: 80-08-0
EINECS: 201-248-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR