Methyltricosanoat

Lieferant: MATREYA

Synonyms: Tricosansäuremethylester

MAYA1187EA 0 CHF
MAYA1187 BTRN1187
Methyltricosanoat
Methyltricosanoat
Formel: C₂₄H₄₈O₂
MW: 368,64 g/mol
MDL Number: MFCD00009349
CAS-Nummer: 2433-97-8
EINECS: 219-420-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR