Methylheptadecanoat

Lieferant: MATREYA

Synonyms: HeptadecansäuremethylesterMargarinsäuremethylesterMethylmargarat

MAYA1019EA 0 CHF
MAYA1019 BTRN1019
Methylheptadecanoat
Methylheptadecanoat
Formel: C₁₈H₃₆O₂
MW: 284,48 g/mol
MDL Number: MFCD00009001
CAS-Nummer: 1731-92-6
EINECS: 217-055-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR