Dapson
Lieferant: EHRENSTORFER GMBH
|
Achtung
|
Synonyms:
DDS
, 4,4'-Diaminodiphenylsulfon
, 4,4'-Sulfonyldianilin
, 4-Aminophenylsulfon
, Bis(4-aminophenyl)sulfon
EHERC11963000EA
77.5
CHF
EHERC11963000
Dapson
Dapson
Formel:
C₁₂H₁₂N₂O₂S MW: 248,31 g/mol |
MDL Number:
MFCD00007887 CAS-Nummer: 80-08-0 EINECS: 201-248-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...