n-Nonacosan
Lieferant: EHRENSTORFER GMBH
EHERC15621000EA
147
CHF
EHERC15621000
n-Nonacosan
n-Nonacosane
Formel:
H₃C(CH₂)₂₇CH₃ MW: 408,8 g/mol Siedepunkt: 440,8 °C (1013 hPa) Melting Pt: 63,7 °C Dichte: 0,8083 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
CAS-Nummer:
630-03-5 EINECS: 211-126-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...