n-Hentriacontan

Lieferant: EHRENSTORFER GMBH

EHERC14085500EA 143 CHF
EHERC14085500
n-Hentriacontan
n-Hentriacontane
Formel: H₃C(CH₂)₂₉CH₃
MW: 436,85 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00048745
CAS-Nummer: 630-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR