1-Butanol ≥99,4% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
31068.AP
31068.K2
31068.M1
31068.APEA
71.9
CHF
31068.AP
31068.K2
31068.M1
1-Butanol ≥99,4% ACS
1-Butanol
Purity: 99.4+%
CAS No.: 71-36-3
Molecular Formula: CH3(CH2)3OH
Formula Weight: 74.12
Flash Point: 35°(95°F)
Physical Form: Liquid
Density: 0.810
Refractive Index: 1.3990
Odour: Alcohol-like
pH: 7
Melting Point: -90 to -89°
Boiling Point: 117-118°
Beilstein Reference: 969148
Merck Reference: 14,1540
UN No.: 1120
MDL No.: MFCD00002964
Formel:
H₃C(CH₂)₂CH₂OH Siedepunkt: 117…118 °C Melting Pt: –90…–89 °C Dichte: 0.810 Flash-Pt: 35 °C (95 °F) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00002964 CAS-Nummer: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3,III REACH: 01-2119484630-38 Merck Index: 14,1540 |