Membranscheibenfilter, Metricel®
Lieferant: Cytiva (Formerly Pall Lab)
Metricel®
516-8313EA 247 CHF
516-8313
Membranscheibenfilter, Metricel®
Filter Membranfilter
Pure PP membrane for filtration of aggressive solvents.
  • High chemical stability
  • High temperature resistance and low extractable levels
  • Inherently hydrophobic
  • Economical alternative to PTFE