Octylacetat
Lieferant: Roth Carl
Synonyms:
Essigsäureoctylester
ROTH5009.1EA
0
CHF
ROTH5009.1
Octylacetat
Octylacetat
Formel:
C₁₀H₂₀O₂ MW: 172,27 g/mol Siedepunkt: 195…199 °C (1013 hPa) Melting Pt: –38,5 °C Dichte: 0,885 g/cm³ (0 °C) |
MDL Number:
MFCD00009562 CAS-Nummer: 112-14-1 EINECS: 203-939-6 |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...