trans-4-Aminocyclohexanol Hydrochlorid
Lieferant: LGC Standards PROMOCHEM
PROOMM0005.06EA
545
CHF
PROOMM0005.06
trans-4-Aminocyclohexanol Hydrochlorid
trans-4-Aminocyclohexanol Hydrochlorid
Formel:
C₆H₁₄ClNO MW: 151,64 g/mol |
MDL Number:
MFCD00012566 CAS-Nummer: 50910-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...