Natriumtriacetoxyborhydrid

Lieferant: Molekula

MOLE10796774-100GEA 0 CHF
MOLE10796774-100G
Natriumtriacetoxyborhydrid
Natriumtriacetoxyborhydrid
Formel: C₆H₁₀BNaO₆
MW: 211,94 g/mol
Melting Pt: 114…118 °C
MDL Number: MFCD00012211
CAS-Nummer: 56553-60-7
UN: 3396
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR