2-Mercaptobenzimidazol

Lieferant: Molekula

Synonyms: Benzimidazol-2-thiol

MOLE30892131-100GEA 0 CHF
MOLE30892131-100G
2-Mercaptobenzimidazol
2-Mercaptobenzimidazole
Formel: C₇H₆N₂S
MW: 150,2 g/mol
Siedepunkt: 376 °C (1013 hPa)
Melting Pt: 154 °C
Dichte: 1,42 g/cm³ (20 °C)
MDL Number: MFCD00466107
CAS-Nummer: 583-39-1
EINECS: 209-502-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR