2-Acetylfuran

Lieferant: Molekula

Synonyms: 2-Furylmethylketon

MOLE14794858-100GEA 0 CHF
MOLE14794858-100G
2-Acetylfuran
2-Acetylfuran
Formel: C₆H₆O₂
MW: 110,11 g/mol
Siedepunkt: 183 °C (1013 hPa)
Melting Pt: 29…33 °C
Dichte: 1,098 g/cm³ (20 °C)
MDL Number: MFCD00003242
CAS-Nummer: 1192-62-7
EINECS: 214-757-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR