1-Butanol 80% ≥80%

Lieferant: EUROCLONE
Gefahr

Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol

EMR067001 EMR067025 EMR067500
EURLEMR067001EA 0 CHF
EURLEMR067001 EURLEMR067025 EURLEMR067500
1-Butanol 80% ≥80%
1-Butanol
Formel: H₃C(CH₂)₂CH₂OH
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002964
CAS-Nummer: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR