(S)-(+)-Carvon, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyms: (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-onD-p-Mentha-1(6),8-dien-2-onD(+)-Carvon

M-Clarity™ Qualitätsstufe = MQ200

8.18410.0025EA 82.1 CHF
8.18410.0025 8.18410.9999
(S)-(+)-Carvon, Sigma-Aldrich®
(S)-(+)-Carvon
Formel: C₁₀H₁₄O
MW: 150,22 g/mol
Siedepunkt: 230 °C (1013 hPa)
Melting Pt: ? 15 °C
Dichte: 0,9584 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00062997
CAS-Nummer: 2244-16-8
EINECS: 218-827-2

Order Now

Spezifikation Testergebnisse

Hill Formula C₁₀H₁₄O
Molar Mass 150.22 g/mol
Boiling point 228 - 230 °C (1013 hPa)
Density 0.96 g/cm³ (20 °C)
Flash point 88 °C DIN 51758
Refractive Index 1.4988 (20 °C, 589 nm)
Solubility 1300 mg/l (18 °C)
LD 50 oral LD50 Rat 3562 mg/kg
Storage class 10 Combustible liquids not in Storage Class 3
WGK WGK 1 slightly water endangering

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR