Bicin

Lieferant: Molekula

Synonyms: N,N-Bis(2-hydroxyethyl)glycin

MOLEM12063561EA 0 CHF
MOLEM12063561
Bicin
Bicin
Formel: C₆H₁₃NO₄
MW: 163,17 g/mol
Melting Pt: 185…192 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004295
CAS-Nummer: 150-25-4
EINECS: 205-755-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR