Propionaldehyd
Lieferant: Roth Carl
|
Gefahr
|
Synonyms:
Propanal
ROTH6836.1EA
0
CHF
ROTH6836.1
Propionaldehyd
Propionaldehyd
Formel:
C₃H₆O MW: 58,08 g/mol Siedepunkt: 48 °C (1013 hPa) Melting Pt: –81 °C Dichte: 0,811 g/cm³ (20 °C) Flash-Pt: –40 °C Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00007020 CAS-Nummer: 123-38-6 EINECS: 204-623-0 UN: 1275 ADR: 3,II Merck Index: 13,07915 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...