tri-Natriumphosphat Dodecahydrat 98,0-102,0% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98.0-102.0%
CAS No.: 10101-89-0
Molecular Formula: Na3O4P.12H2O
Formula Weight: 380.12 (163.94anhy)
Physical Form: Crystalline
Density: 1.62
Appearance: White
Odour: Odourless
pH: 12
Melting Point: ca 75° dec.
Solubility: Soluble in water
Merck Reference: 14,8662
UN No.: 3262
MDL No.: MFCD00149198
Formel:
Na₃PO₄·12H₂O Melting Pt: ∼75 °C (Zersetzung) Dichte: 1,62 Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00149198 CAS-Nummer: 10101-89-0 EINECS: 231-509-8 UN: 3262 ADR: 8,III REACH: 01-2119489800-32-xxxx Merck Index: 14,8662 |