Schwefelsäure 2.5 mol/l (5 N) in wässriger Lösung, Titripur® volumetrische Lösung, Supelco®
Lieferant: Merck
Gefahr
M-Clarity™ Qualitätsstufe = MQ100
4.80364.1000 4.80364.9025
4.80364.1000EA 42.1 CHF
4.80364.1000 4.80364.9025
Schwefelsäure 2.5 mol/l (5 N) in wässriger Lösung, Titripur® volumetrische Lösung, Supelco®
Schwefelsäure
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formel: H₂SO₄
MW: 98.08 g/mol
Siedepunkt: 330
Melting Pt: 10.38
Dichte: 1.146 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00064589
CAS-Nummer: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II