CAPS (3-(Cyclohexylamino)propansulfonsäure)

Lieferant: Merck

1.06195.0500EA 0 CHF
1.06195.0500
CAPS (3-(Cyclohexylamino)propansulfonsäure)
CAPS (3-(Cyclohexylamino)propansulfonsäure)
Formel: C₆H₁₁NH(CH₂)₃SO₃H
MW: 221,32 g/mol
Melting Pt: 324 °C
MDL Number: MFCD00003837
CAS-Nummer: 1135-40-6
Merck Index: 13,01773

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR