CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Lieferant: Molekula
MOLE11091556-500GEA
0
CHF
MOLE11091556-500G
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Formel:
C₉H₁₉NO₄S MW: 237,32 g/mol Melting Pt: 300 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00041778 CAS-Nummer: 73463-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...