CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)

Lieferant: Molekula

MOLEM12546873EA 0 CHF
MOLEM12546873
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Formel: C₉H₁₉NO₄S
MW: 237,32 g/mol
Melting Pt: 300 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00041778
CAS-Nummer: 73463-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR