CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)

Lieferant: Merck

1.06186.0100EA 0 CHF
1.06186.0100
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Formel: C₉H₁₉NO₄S
MW: 237,32 g/mol
Melting Pt: 270 - 274 °C
MDL Number: MFCD00041778
CAS-Nummer: 73463-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR