ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2), Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyms: Nadide phosphate disodium salt, Triphosphopyridine nucleotide, disodium salt, NADP-Na2, β-NADP-Na2, NADP disodium salt, Nicotinamide-adenine dinucleotide phosphate disodium salt, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium, disodium salt, β-NADH phosphate disodium salt
481972-1 481972-100
481972-1EA 1140 CHF
481972-1 481972-100
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2), Millipore®
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Formel: C₂₁H₂₆N₇Na₂O₁₇P₃
MW: 787,37 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00065390
CAS-Nummer: 24292-60-2
EINECS: 246-129-8