Disulfinblau

Lieferant: Molekula

Synonyms: Acid Blue1Hydrogen[4-[4-(diethylamino)-2',4'-disulfonatobenzhydryliden]cyclohexa-2,5-dien-1-yliden]diethylammonium, NatriumsalzPatentblau VF Natriumsalz

MOLE10362000-25GEA 0 CHF
MOLE10362000-25G
Disulfinblau
Disulfinblau
Formel: C₂₇H₃₁N₂NaO₆S₂
MW: 566,67 g/mol
Melting Pt: 290 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00012119
CAS-Nummer: 129-17-9
EINECS: 204-934-1
Merck Index: 12,09161

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR