Ecoscint A zur Szintillation
Lieferant: NATIONAL DIAGNOSTICS
Ecoscint A displays exceptional sample holding capability while still delivering high efficiency, easily accommodating up to 40% sample while maintaining a single liquid phase. Furthermore, Ecoscint A has exceptional resistance to photoluminescence and chemiluminescence.
- The Ideal Universal Scintillation Cocktail
- All Around Performance
- High Efficiency with High Sample Holding
- DOC elimination of >70% at 10 days, Reduced Toxicity Solvent
- High Flash Point (>150°C)
The high flash point, low odor and low toxicity of Ecoscint A make it perfect for benchtop work, as it is not necessary to use or store under a fume hood.
Ecoscint A maintains its high sample hold capacity for a broad range of solution types, delivering high efficiency counting for many different types of samples.
UN:
1992 ADR: 3,III |