Brandschutzzeichen
Lieferant: KROSCHKE
Z5887 W17027 W17033 W2395 Z5908 Z5905
116-1140EA 291 CHF
116-1140 116-2016 116-2019 116-2024 116-2026 116-2027
Brandschutzzeichen
Warn- und Verbotszeichen
These self-adhesive, PVC film signs are used to indicate fire equipment. According to EC Directive 92/58/EEC and related national legislation on safety and/or health signs and also according to to VBG 125 (a regulation of the German employer's liability insurance association), signboards/signs for fire equipment must be permanently fitted and clearly recognisable at all times.
  • Suitable for use inside and outside
  • Visible from a distance of ∼12 metres
  • Long-lasting persistent luminescence material available
Zertifizierungen: VBG 125 (now known as BGV A8).More Product Information