IsoTherm-System®
Lieferant: Eppendorf
IsoTherm-System®
479-1142EA 164 CHF
479-1142 479-1131 479-1141 479-1133 479-1110 479-1132
IsoTherm-System®
Boxen Kühl- und Gefrierboxen, Kühl- und Gefrierbeutel
IsoTherm benchtop cooler.
IsoTherm cooler eliminates the messiness and contamination risk of ice buckets while protecting temperature-sensitive samples and enzymes during use on the benchtop. The system can maintain 0 °C up to 6 hours and −21 °C for 3 hours. It utilizes a modular rack system numbered for easy sample storage and identification. Each autoclavable IsoRack holds twenty-four 0,5 ml or 1,5 to 2,0 ml tubes, withstands temperatures down to −200 °C , floats in a water bath while thawing or incubating samples, and can be centrifuged in a microplate rotor.
Starter sets are a complete system consisting of one 0 °C IsoPack cold pack, one −21 °C IsoPack cold pack, one transparent IsoRack, and one IsoSafe insulated transport rack. IsoPack and IsoRack sets may also be purchased separately.