Kupfer(II)hydroxid

Lieferant: EHRENSTORFER GMBH

Synonyms: Kupferdihydroxid

EHERC11698000EA 94.7 CHF
EHERC11698000
Kupfer(II)hydroxid
Copper(II) hydroxide
Formel: H₂CuO₂
MW: 97,56 g/mol
Dichte: 3,37 g/cm³ (20 °C)
MDL Number: MFCD00010968
CAS-Nummer: 20427-59-2
EINECS: 243-815-9
UN: 3288
ADR: 6.1,III
Merck Index: 12,02709

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR