Anthralin
Lieferant: EHRENSTORFER GMBH
Synonyms:
Dithranol
, 1,8,9-Trihydroxyanthracen
, 1,8,9-Anthracentriol
, 1,8-Dihydroxyanthracen-9(10H)-on
, Anthranol
EHERC13013900EA
102
CHF
EHERC13013900
Anthralin
Anthralin
Formel:
C₁₄H₁₀O₃ MW: 226,23 g/mol Siedepunkt: 400 °C (1013 hPa) Melting Pt: 193,5…195 °C Dichte: 1,433 g/cm³ (20 °C) |
MDL Number:
MFCD00053409 CAS-Nummer: 1143-38-0 EINECS: 214-538-0 Merck Index: 12,00723 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...