EGTA (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäure)

Lieferant: EUROCLONE

Synonyms: EGTA, Ethylenebis(oxyethylenenitrilo)tetra(acetic acid), Kestranal® FE PA, Idranal® VI, Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, Ethylenebis(oxyethylenenitrilo)tetraacetic acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid, Titriplex® VI

EMR132025 EMR132100
EURLEMR132025EA 0 CHF
EURLEMR132025 EURLEMR132100
EGTA (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäure)
EGTA (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäure)
Formel: C₁₄H₂₄N₂O₁₀
MW: 380.35 g/mol
Melting Pt: 240…244 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004291
CAS-Nummer: 67-42-5
EINECS: 200-651-2
Merck Index: 13,03560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR