Coomassie Brillantblau G 250

Lieferant: EUROCLONE

EMR114010G EMR114100G
EURLEMR114010GEA 0 CHF
EURLEMR114010G EURLEMR114100G
Coomassie Brillantblau G 250
Coomassie Brillantblau G 250
Formel: C₄₇H₄₈N₃NaO₇S₂
Dichte: >1 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00078482
CAS-Nummer: 6104-58-1
EINECS: 228-058-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR