N,N-Diethyl-m-toluamid

Lieferant: Molekula
Achtung

Synonyms: Deetm-TolylsäurediethylamidN,N-Diethyl-3-methylbenzamidDiethyl-m-toluamid

MOLE27939809-500GEA 57.89 CHF
MOLE27939809-500G
N,N-Diethyl-m-toluamid
N,N-Diethyl-m-toluamid
Formel: C₁₂H₁₇NO
MW: 191,27 g/mol
Siedepunkt: 298 °C (1013 hPa)
Melting Pt: –45 °C
Dichte: 0,996 g/cm³ (20 °C)
Flash-Pt: 140 °C
MDL Number: MFCD00009046
CAS-Nummer: 134-62-3
EINECS: 205-149-7
Merck Index: 13,02876

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR