IPTG (Isopropyl-β-D-thiogalactopyranosid)
Lieferant: EUROCLONE
Synonyms:
IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
EMR066001
EMR066005
EMR066025
EMR066050
EMR066100
EURLEMR066001EA
0
CHF
EURLEMR066001
EURLEMR066005
EURLEMR066025
EURLEMR066050
EURLEMR066100
IPTG (Isopropyl-β-D-thiogalactopyranosid)
IPTG (Isopropyl-β-D-thiogalactopyranosid)
Formel:
C₉H₁₈O₅S Siedepunkt: 402 °C (1013 hPa) Melting Pt: 109.5…110.5 °C Lager Temperatur: Gefrierschrank |
MDL Number:
MFCD00063273 CAS-Nummer: 367-93-1 EINECS: 206-703-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...