Cefuroxime axetil

Lieferant: Thermo Fisher Scientific

468670010 468672500
468670010.EA 173 CHF
468670010. 468672500.
Cefuroxime axetil
Cefuroxime axetil
Formel: C₂₀H₂₂N₄O₁₀S
MDL Number: MFCD00864991
CAS-Nummer: 64544-07-6

Order Now

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥96.0 % (sum of diastereoisomers)
Water ≤1.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR