Silbernitrat 0,1 mol/L
Lieferant: CPACHEM
|
Achtung
|
Synonyms:
Höllenstein
ACSDCH8006024.1LEA
433
CHF
ACSDCH8006024.1L
Silbernitrat 0,1 mol/L
Silbernitrat
Zertifizierungen: ISO 9001 certified.
Bestellinformation: Shelf life on ship date: 24 months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Formel:
AgNO₃ MW: 169,87 g/mol Dichte: 4,35 g/cm³ |
MDL Number:
MFCD00003414 CAS-Nummer: 7761-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...