X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)

Lieferant: EUROCLONE

Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

EMR047001 EMR047005 EMR047010 EMR047250
EURLEMR047001EA 0 CHF
EURLEMR047001 EURLEMR047005 EURLEMR047010 EURLEMR047250
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
X-Gal (5-Brom-4-chlor-3-indolyl-ß-D-galactopyranosid)
Formel: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Melting Pt: 237…239 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00005666
CAS-Nummer: 7240-90-6
EINECS: 230-640-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR