Alternariol

Lieferant: LGC Standards PROMOCHEM
Gefahr

PROOTRCA575760-5MGEA 672 CHF
PROOTRCA575760-5MG
Alternariol
Alternariol
Formel: C₁₄H₁₀O₅
MW: 258,226 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00133068
CAS-Nummer: 641-38-3
EINECS: 636-339-1
UN: 3462
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR