o-Kresol
Lieferant: EHRENSTORFER GMBH
|
Gefahr
|
Synonyms:
2-Methylphenol
, 2-Hydroxytoluol
, 1-Hydroxy-2-methylbenzol
EHERCA15140200EA
0
CHF
EHERCA15140200
o-Kresol
o-Kresol
Formel:
C₇H₈O MW: 108,14 g/mol Siedepunkt: 31 °C Flash-Pt: 81 °C Lager Temperatur: Room Temperature |
MDL Number:
MFCD00002226 CAS-Nummer: 95-48-7 UN: 2076 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...