L(-)-Prolin *copeia

Lieferant: CUSTOM MADE CHEMICALS

2031-100G 2031-5KG
CMPP2031-100GEA 0 CHF
CMPP2031-100G CMPP2031-5KG
L(-)-Prolin *copeia
L(-)-Prolin
Formel: C₅H₉NO₂
Lager Temperatur: Room Temperature
MDL Number: MFCD00064318
CAS-Nummer: 147-85-3
EINECS: 205-702-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR