Petrolatum, weiß

Lieferant: USP

1514911.EA 387 CHF
1514911.
Petrolatum, weiß
Petrolatum
Melting Pt: 35,..80 °C (95...176 °F)
MDL Number: MFCD00147839
CAS-Nummer: 8009-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR