2-Hydroxy-5-methoxyisoindoline-1,3-dione ≥98,0% (durch HPLC, titrimetrische Analyse)
Lieferant: TCI EUROPE
TCIAM3572-5GEA
0
CHF
TCIAM3572-5G
2-Hydroxy-5-methoxyisoindoline-1,3-dione ≥98,0% (durch HPLC, titrimetrische Analyse)
2-Hydroxy-5-methoxyisoindoline-1,3-dione
Formel:
C₉H₇NO₄ MW: 193,16 g/mol Melting Pt: 220 °C Lager Temperatur: Room Temperature |
CAS-Nummer:
214754-50-4 |
Spezifikation Testergebnisse
Appearance | White to Light yellow powder to crystal |
Purity(HPLC) | min. 98.0 area% |
Purity(Neutralization titration) | min. 98.0 % |
Melting point | 218.0 to 222.0 °C |
NMR | confirm to structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...