2-Hydroxy-5-methylisoindoline-1,3-dione ≥98,0% (durch HPLC, titrimetrische Analyse)

Lieferant: TCI EUROPE

TCIAM3571-5GEA 0 CHF
TCIAM3571-5G
2-Hydroxy-5-methylisoindoline-1,3-dione ≥98,0% (durch HPLC, titrimetrische Analyse)
2-Hydroxy-5-methylisoindoline-1,3-dione
Formel: C₉H₇NO₃
MW: 177,16 g/mol
Melting Pt: 201 °C
Lager Temperatur: Room Temperature
CAS-Nummer: 173962-59-9

Order Now

Spezifikation Testergebnisse

Appearance White to Light yellow powder to crystal
Purity(HPLC) min. 98.0 area%
Purity(Neutralisation titration) min. 98.0 %
Melting point 199.0 to 203.0 °C
NMR confirm to structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR