Anti-KL Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP1-76511EA 0 CHF
NOVUNBP1-76511
Anti-KL Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal Klotho Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: KL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR