Anti-IgG1 Monoclonal Antibody [clone: 001]

Lieferant: USP
1445539.EA 1380 CHF
1445539.
Anti-IgG1 Monoclonal Antibody [clone: 001]
Antikörper
Type:
Antigen: IgG1
Clonality: Monoclonal
Clone: 001
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR