Thiamin Hydrochlorid, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyms: Thiaminchlorid HydrochloridThiaminiumchloridantineuritisches Vitamin HydrochloridAntiberiberifaktor HydrochloridAneurin HydrochloridVitamin B1 Hydrochlorid

5871-100EA 153 CHF
5871-100
Thiamin Hydrochlorid, Millipore®
Thiamin Hydrochlorid
Formel: C₁₂H₁₈Cl₂N₄OS
MW: 337,27 g/mol
Melting Pt: 252 °C
Dichte: 1,401 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00012780
CAS-Nummer: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR