1-Dodecansulfonsäure Natriumsalz

Lieferant: CPACHEM

Synonyms: NatriumlaurylsulfonatNatriumdodecylsulfonat

ACSDSB48924.100MGEA 182 CHF
ACSDSB48924.100MG
1-Dodecansulfonsäure Natriumsalz
1-Dodecansulfonsäure Natriumsalz
Formel: C₁₂H₂₅NaO₃S
MW: 272,38 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD00007527
CAS-Nummer: 2386-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR